Anti-SLC2A10 Rabbit Polyclonal Antibody

Supplier: Prosci

48-773
10076-016EA 1011.8 CAD
10076-016
Anti-SLC2A10 Rabbit Polyclonal Antibody
Antibodies
SLC2A10 antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC2A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR