Anti-NPM Rabbit Polyclonal Antibody

Supplier: Prosci

48-792
10076-044EA 955.07 CAD
10076-044
Anti-NPM Rabbit Polyclonal Antibody
Antibodies
NPM1 is a ubiquitously expressed nucleolar protein that shuttles between the nucleus and cytoplasm. It is implicated in multiple functions, including ribosomal protein assembly and transport, control of centrosome duplication, and regulation of the tumor suppressor ARF. NPM1 mutations that relocalize NPM1 from the nucleus into the cytoplasm are associated with development of acute myeloid leukemia.

Nucleophosmin antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol. BSA is present at 1 mg/mL final concentration (USA origin).

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR