Anti-SYK Mouse Monoclonal Antibody [clone: [SYK-01]]

Supplier: Prosci

48-803
10076-058EA 1011.8 CAD
10076-058
Anti-SYK Mouse Monoclonal Antibody [clone: [SYK-01]]
Antibodies
SYK antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: SYK
Clonality: Monoclonal
Clone: [SYK-01]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR