Order Entry
ContactUsLinkComponent
Anti-NFKB1 Rabbit Polyclonal Antibody
Anti-NFKB1 Rabbit Polyclonal Antibody
Catalog #: 10074-426
Supplier:  Avantor
Anti-NFKB1 Rabbit Polyclonal Antibody
Catalog #: 10074-426
Supplier:  Avantor
Discontinued
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
  • Clonality:
    Polyclonal
  • Gene ID:
    P19838
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Antigen symbol:
    NFKB1
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    NFKB1
  • Storage buffer:
    Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 0.2% gelatin.
  • Storage temperature:
    Store NFKB1 antibody at 4 °C, Avoid Freezing
  • Shipping temperature:
    4 °C
  • Immunogen:
    NFKB1 antibody was raised against a peptide located in the C Terminus of NFKB1 (Human).
  • Tested applications:
    Immunoaffinity Chromatography
  • Purification:
    Immunoaffinity Chromatography
  • Cat. no.:
    10074-426

Specifications

About this item

NFKB1 antibody can be used in ELISA, Western Blot starting at 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and flow cytometry.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 0.2% gelatin.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: NFKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Customers Who Bought This Also Bought