Anti-MAP2K2 Rabbit Polyclonal Antibody

48-876
10076-074EA 811.46 CAD
10076-074
Anti-MAP2K2 Rabbit Polyclonal Antibody
MEK2, a MAP2K type protein kinase, is a member of the Ras MAPK pathway. This pathway modulates the activity of transcription factors, structural proteins and other protein kinases, leading to biological responses such as cell proliferation and differentiation. MEK2 contains a proline-rich segment that is recognized by the scaffold protein MEK binding partner-1 in vitro but not in vivo. Similar to MEK1, MEK2 is phosphorylated by several kinases, including Raf1, c-Mos and MEKK1. Activated MEK2 associates with and phosphorylates MAP kinases ERK1 and 2. The inhibition or degradation of this kinase is found to be involved in the pathogenesis of Yersinia and anthrax.

MAP2K2 antibody can be used in ELISA starting at 2.5 ?g/mL, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR