Anti-MAP2K4 Rabbit Polyclonal Antibody

48-877
10076-076EA 811.46 CAD
10076-076
Anti-MAP2K4 Rabbit Polyclonal Antibody
MKK4, a MAP2K type protein kinase, is a member of the stress-activated protein kinase (SAPK) pathways which convey pro-apoptotic signals. MKK4 is activated by MEKK1 or MEKK2, and to a lesser extent by GCK or ASK1. MKK4 phosphorylates and activates JNK1, JNK2 and p38. MKK4 deficiency in mouse embryos led to lethality after embryonic day 12.5 and was associated with abnormal liver development. Additional studies have linked MKK4 to tumor suppression.

MAP2K4 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR