Anti-TRHR Rabbit Polyclonal Antibody

Supplier: Prosci
48-927
10076-116EA 955.07 CAD
10076-116
Anti-TRHR Rabbit Polyclonal Antibody
Antibodies
The thyrotropin-releasing hormone receptor (TRH-R), a Releasing Hormone Receptor, is activated by thyrotropin-releasing hormone. TRH-R may play a role in the etiopathogenesis of essential hypertension.

  • TRHR antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Dog, Human, Monkey, Mouse, Opossum, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR