Anti-SRI Rabbit Polyclonal Antibody

Supplier: Prosci

48-932
10076-126EA 1047.24 CAD
10076-126
Anti-SRI Rabbit Polyclonal Antibody
Antibodies
Sorcin antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SRI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Chicken, Dog, Xenopus, Horse, Human, Monkey, Mouse, Opossum, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR