Anti-MTSS1 Rabbit Polyclonal Antibody

Supplier: Prosci

48-957
10078-400EA 1011.8 CAD
10078-400
Anti-MTSS1 Rabbit Polyclonal Antibody
Antibodies
Metastasis Suppressor 1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTSS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chimpanzee, Dog, Horse, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR