Anti-AQP2 Rabbit Polyclonal Antibody

Supplier: Prosci

48-961
10078-408EA 1011.8 CAD
10078-408
Anti-AQP2 Rabbit Polyclonal Antibody
Antibodies
Aquaporin 2 antibody can be used in Western Blot starting at 5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AQP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR