Anti-DNM1 Rabbit Polyclonal Antibody

Supplier: Prosci
48-966
10078-418EA 955.07 CAD
10078-418
Anti-DNM1 Rabbit Polyclonal Antibody
Antibodies
Dynamin 1 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR