Anti-CD59 Mouse Monoclonal Antibody [clone: [MEM-43/5]]

48-992
10078-470EA 811.46 CAD
10078-470
Anti-CD59 Mouse Monoclonal Antibody [clone: [MEM-43/5]]
CD59 antibody can be used in immunohistochemistry starting at 15 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: [MEM-43/5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR