Anti-B2M Mouse Monoclonal Antibody [clone: [B2M-02]]

48-997
10076-152EA 811.46 CAD
10076-152
Anti-B2M Mouse Monoclonal Antibody [clone: [B2M-02]]
beta2M is a 12 kDa Ig like glycoprotein expressed on lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells and epithelial cells. It is absent on erythrocytes. Beta 2-microglobulin is the low molecular weight component of HLA antigens and as such is involved in the presentation of endogenous antigens, activation of T lymphocytes and functional emergence of natural killer cells. It is found in the serum of normal individuals and in the urine in elevated amounts in patients with Wilson disease, cadmium poisoning and other conditions leading renal tubular dysfunction. ESTs have been isolated from libraries generated using virtually every tissue in the body.

B2M antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: [B2M-02]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR