Anti-APOA1 Mouse Monoclonal Antibody [clone: [057-10029]]

Supplier: ProSci

49-003
10076-160EA 1011.8 CAD
10076-160
Anti-APOA1 Mouse Monoclonal Antibody [clone: [057-10029]]
Antibodies
APOA1 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOA1
Clonality: Monoclonal
Clone: [057-10029]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR