Anti-CD34 Mouse Monoclonal Antibody

Supplier: ProSci

49-041
10078-528EA 1011.8 CAD
10078-528
Anti-CD34 Mouse Monoclonal Antibody
Antibodies
CD34 antibody can be used in immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1:25 - 1:100.

PBS, 0.1% sodium azide and a stabilizing agent.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR