Anti-ACTB Mouse Monoclonal Antibody [clone: [4E8H3]]

49-048
10076-180EA 811.46 CAD
10076-180
Anti-ACTB Mouse Monoclonal Antibody [clone: [4E8H3]]
Beta-actin antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: [4E8H3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR