Anti-ICAM1 Mouse Monoclonal Antibody [clone: [15.2]]

Supplier: ProSci
49-050
10076-184EA 1047.24 CAD
10076-184
Anti-ICAM1 Mouse Monoclonal Antibody [clone: [15.2]]
Antibodies
ICAM1 antibody can be used in Western Blot starting at 0.5 - 4 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: [15.2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR