Anti-BRCC3 Mouse Monoclonal Antibody [clone: [AT3B1]]

Supplier: ProSci

49-077
10078-574EA 1011.8 CAD
10078-574
Anti-BRCC3 Mouse Monoclonal Antibody [clone: [AT3B1]]
Antibodies
BRCC36 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BRCC3
Clonality: Monoclonal
Clone: [AT3B1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR