Anti-DPYSL2 Mouse Monoclonal Antibody [clone: [33CT11.40.3]]

Supplier: ProSci

49-080
10078-580EA 1047.24 CAD
10078-580
Anti-DPYSL2 Mouse Monoclonal Antibody [clone: [33CT11.40.3]]
Antibodies
CRMP2 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and flow cytometry.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DPYSL2
Clonality: Monoclonal
Clone: [33CT11.40.3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR