Anti-Troponin T type 2 Mouse Monoclonal Antibody

49-085
10078-590EA 811.46 CAD
10078-590
Anti-Troponin T type 2 Mouse Monoclonal Antibody
CTnT antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.2% BSA, 0.09% sodium azide, before the addition of glycerol to 40%.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Bovine, Chicken, Dog, Gorilla, Horse, Human, Monkey, Mouse, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR