Anti-SELP Mouse Monoclonal Antibody [clone: [Psel.KO.2.5]]

Supplier: ProSci

49-086
10078-592EA 1011.8 CAD
10078-592
Anti-SELP Mouse Monoclonal Antibody [clone: [Psel.KO.2.5]]
Antibodies
CD62P antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: [Psel.KO.2.5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR