Anti-CASP7 Mouse Monoclonal Antibody [clone: [7-1-11]]

Supplier: ProSci

49-092
10076-204EA 1047.24 CAD
10076-204
Anti-CASP7 Mouse Monoclonal Antibody [clone: [7-1-11]]
Antibodies
Caspase 7 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 ?g/mL.

PBS (10 mM sodium phosphate, pH 7.2, 50 mM NaCl, 0.1 mM PMSF)

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CASP7
Clonality: Monoclonal
Clone: [7-1-11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR