Anti-MAP2 Mouse Monoclonal Antibody [clone: [MT-07]]

49-110
10076-240EA 811.46 CAD
10076-240
Anti-MAP2 Mouse Monoclonal Antibody [clone: [MT-07]]
MAP2 antibody can be used in ELISA, and immunohistochemistry starting at 15 ?g/mL.

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: [MT-07]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR