Anti-CASR Rabbit Polyclonal Antibody

49-156
10078-652EA 620.52 CAD
10078-652
Anti-CASR Rabbit Polyclonal Antibody
CASR, a Calcium-Sensing Receptor, functions as a sensor in parathyroid and kidney to determine extracellular calcium concentration and to maintain calcium homeostasis. This receptor regulates both secretion of parathyroid hormone and excretion of urinary calcium. Mutations that inactivate CASR cause familial hypocalciuric hypercalcemia, which is also correlated with hyperparathyroidism. On the other hand, mutations that activate CASR cause autosomal dominant hypocalcemia. Hjalm et al. (2001) and Awata et al. (2001) have identified the binding of filamin-1 to the C-terminal tail of CASR, which may contribute to its localization in caveolae, link it to the actin-based cytoskeleton, and participate in CASR-mediated activation of MAPK. Multiple isoforms for CASR are produced by alternative splicing.

Calcium-Sensing Receptor antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.2% BSA, 0.05% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: CASR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR