Anti-ADD1 Rabbit Polyclonal Antibody

49-187
10076-268EA 776.14 CAD
10076-268
Anti-ADD1 Rabbit Polyclonal Antibody
ADD1 antibody can be used in Western Blot starting at 0.5 - 1 ?g/mL, and immunofluorescence.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR