Anti-CD40 Mouse Monoclonal Antibody

49-202
10078-672EA 623.55 CAD
10078-672
Anti-CD40 Mouse Monoclonal Antibody
CD40 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2, 0.09% sodium azide.

Store CD40 antibody undiluted at 4 °C.

Type:
Antigen: CD40
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR