Anti-MUTYH Rabbit Polyclonal Antibody

Supplier: ProSci
49-220
10078-698EA 955.07 CAD
10078-698
Anti-MUTYH Rabbit Polyclonal Antibody
Antibodies
MYH is the eukaryotic homolog to the E. coli protein MutY. It was first identified in HeLa cells and nicks DNA replication errors, specifically A/G mismatches.

MUTYH antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUTYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR