Anti-MYD88 Rabbit Polyclonal Antibody

Supplier: ProSci

49-221
10078-700EA 954.81 CAD
10078-700
Anti-MYD88 Rabbit Polyclonal Antibody
Antibodies
MyD88, a protein involved in interleukin-1 (IL-1) mediated signalling was originally isolated as myeloid differentiation primary response gene. MyD88 possesses a N-terminal death domain similar to cytoplasmic segments of TNF receptor 1, Fas, and C-terminal region related to IL-1 and Toll receptors. Overexpression of MyD88 induces activation the c-Jun N-terminal kinase and NF-κB through its death domain (1,2). The C-terminus of MyD88 interacts with the IL-1 receptor and blocks NF-κB activation induced by IL-1, but not by NF-κB. The pro-inflammatory cytokine IL-1 induced cellular response requires IL-1 receptor complex including IL-1RI and IL-1RAcP. Recently, MyD88 was identified as an adapter molecule in the IL-1 signaling pathway. MyD88 associates with and recruits IRAK to the IL-1 receptor complex in response to IL-1 treatment and dominant negative form of MyD88 attenuates IL-1R-mediated NF-κB activation. MyD88 is also employed as a regulator molecule by IL-18 receptor and human Toll receptor, which are members in the Toll/IL-1R family of receptors. Targeted disruption of the MyD88 gene results in lose of cellular responses to IL-1 and IL-18, and MyD88-deficient mice lack responses to bacterial product LPS that employs Toll-like receptors 2 and 4 (TLR2 and TLR4) as the signaling receptors. MyD88 is a general adapter protein for the Toll/IL-1R family of receptors and plays an important role in the inflammatory response induced by cytokines IL-1 and IL-18 and endotoxin. MyD88 gene is expressed in many tissues.

MYD88 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MYD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR