Anti-LMNA Rabbit Polyclonal Antibody

Supplier: ProSci
49-252
10078-724EA 955.07 CAD
10078-724
Anti-LMNA Rabbit Polyclonal Antibody
Antibodies
Lamin A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 20 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LMNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR