Anti-ATG7 Rabbit Polyclonal Antibody

Supplier: ProSci

49-385
10078-868EA 1067.97 CAD
10078-868
Anti-ATG7 Rabbit Polyclonal Antibody
Antibodies
Apg7 is the human gene product similar in homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae Apg7. In the yeast, the protein appears to be required for fusion of peroxisomal and vaculuolar membranes. The protein shows homlogy to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.

ATG7 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR