Anti-CCNL2 Rabbit Polyclonal Antibody

49-391
10076-368EA 620.52 CAD
10076-368
Anti-CCNL2 Rabbit Polyclonal Antibody
Cyclin L (also referred to as CCNL) is encoded by two highly related genes Cyclin L1 and Cyclin L2 (CCNL1 and CCNL2, respectively). Cyclin L2 can be alternatively spliced to produce two isoforms (known as 1 and 2 or alpha and beta). The Cyclin L2 gene is located approximately 100-200 kb from the PITSLRE/CDK11 protein kinase locus in human (1p36) and in a syngenetic location on mouse chromosome 4q. Cyclin L2 is ubiquitous, expressed at much higher levels than Cyclin L1, and thus is likely the major partner for the CDK11p110 protein kinase.

Cyclin L2 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCNL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR