Anti-DAXX Rabbit Polyclonal Antibody

Supplier: ProSci
49-419
10078-918EA 1101.77 CAD
10078-918
Anti-DAXX Rabbit Polyclonal Antibody
Antibodies
DAXX, the Death domain associated protein 6 (also named Fas death domain associated protein, ETS1-associated protein 1 or EAP1), functions in part as a transcriptional co-repressor through its interaction with a number of nuclear, DNA-associated proteins, such as histone deacetylase II, core histones H2A, H2B, H3 and H4, and Dek, a chromatin- associated protein reported to change the topology of DNA in chromatin in vitro. DAXX also interacts with p73, and is involved in the regulation of the cell cycle and apoptosis through transcriptional regulation of p53 and possibly its family members.

DAXX antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DAXX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR