Anti-IRAK3 Rabbit Polyclonal Antibody

49-445
10078-960EA 623.55 CAD
10078-960
Anti-IRAK3 Rabbit Polyclonal Antibody
IRAK-M (Interleukin-1 receptor-associated kinase M). Interleukin-1 (IL-1) and lipopolysaccharide (LPS) induce cellular response through IL-1 receptor (IL-1R) and Toll like receptors (TLR). Through IL-1/Toll receptors, IL-1 receptor-associated kinase (IRAK and IRAK2) mediates the activation of NF-κB (1,2); a pivotal transcription factor mediating inflammatory and immune response. A novel member in the IRAK/Pelle family was recently identified and designated IRAK-M. IRAKs associate with IL-1/Toll receptors after IL-1 or LPS stimulation and the dominant negative mutants of IRAKs inhibit IL-1 or LPS induced NF-κB activation. Members of the IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses to cytokine IL-1 and LPS.

IRAK3 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IRAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR