Anti-NOTCH2 Rabbit Polyclonal Antibody

Supplier: ProSci

49-458
10078-984EA 1046.96 CAD
10078-984
Anti-NOTCH2 Rabbit Polyclonal Antibody
Antibodies
Notch is synthesized in the endoplasmic reticulum as an inactive form which is proteolytically cleaved by a furin-like convertase (S1 cleavage) in the trans-golgi network before it reaches the plasma membrane to yield an active, ligand-accessible form. Cleavage results in a C-terminal fragment N(TM) and a N-terminal fragment N(EC). Following ligand binding, it is cleaved (S2 cleavage) by TNF-alpha converting enzyme (TACE) to yield a membrane-associated intermediate fragment called Notch extracellular truncation (NEXT). This fragment is then cleaved by presenilin-dependent gamma-secretase (S3 cleavage) to release the intracellular domain (NICD) from the membrane. Notch functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Upon ligand activation through the released notch intracellular domain (NICD) it forms a transcriptional activator complex with RBP-J kappa and activates genes of the enhancer of split locus. Affects the implementation of differentiation, proliferation and apoptotic programs.

NOTCH2 antibody can be used in Western Blot starting at 0.5 - 4 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

Sterile Filtration

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR