Anti-RFFL Rabbit Polyclonal Antibody

Supplier: ProSci

49-478
10079-010EA 1011.8 CAD
10079-010
Anti-RFFL Rabbit Polyclonal Antibody
Antibodies
RFFL, also known as Rififylin, RING finger and FYVE-like domain-containing protein 1, FYVE-RING finger protein, Sakura, Fring, Caspases-8 and -10-associated RING finger protein 2, CARP-2, Caspase regulator CARP2, RING finger protein 189 and RING finger protein 34-like, is a novel modulator of NF-κB activation. RFFL possesses E3 ubiquitin protein ligase activity and has been shown to regulate the levels of CASP8 and CASP10 by targeting them for proteasomal degradation. RFFL also possesses anti-apoptotic activity and may bind phosphatidylinositol phosphates. RFFL is a membrane bound cytoplasmic protein that is expressed ubiquitously. RFFL can be detected in spleen, thymus, prostate, testis, ovary, small intestine, colon and peripheral blood leukocytes and is rapidly degraded after stimulation with TNFSF10, and probably by caspases. Multiple transcript variants have been detected for this protein.

RFFL antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

Protein A Column

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RFFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR