Anti-TG Mouse Monoclonal Antibody

49-491
10079-028EA 736.85 CAD
10079-028
Anti-TG Mouse Monoclonal Antibody
Thyroglobulin (Tg) is synthesized by the follicular epithelial cells of the thyroid and secreted from the thyroid gland with the stimulation of TSH and/or thyroid stimulating immunoglobulins. Thyroglobulin is a prognostic marker for Graves` disease. Thyroglobulin antibody has been useful in the positive identification of thyroid carcinomas of the papillary and follicular types.

Thyroglobulin antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Protein A Column

0.02 M potassium phosphate, 0.5 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR