Anti-SPANXC Rabbit Polyclonal Antibody

Supplier: ProSci

49-505
10079-050EA 1011.8 CAD
10079-050
Anti-SPANXC Rabbit Polyclonal Antibody
Antibodies
Human Sperm Proteins Associated with the Nucleus on X-chromosome (SPANX) are relatively low molecular weight cytoplasmic proteins found in testis and sperm. Their expression in other tissues indicates malignancies. Family members are observed as proteins that range from 15 to 20 kDa.

SPANXC antibody can be used in Western Blot at 0.5 - 4 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation starting at 10 - 20 ?g/mL.

Protein A Column

PBS, pH 7.2, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR