Anti-S1PR4 Rabbit Polyclonal Antibody

Supplier: ProSci

49-513
10079-062EA 1011.8 CAD
10079-062
Anti-S1PR4 Rabbit Polyclonal Antibody
Antibodies
EDG6, a Lysophospholipid/Lysosphingolipid Receptor, may regulate lymphocyte cell signaling. EDG6 is activated by sphingosine-1-phosphate.

EDG6 antibody can be used in ELISA starting at 1:5000 - 1:20000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 2.5 ?g/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S1PR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR