Anti-DLL1 Rabbit Polyclonal Antibody

Supplier: ProSci

49-592
10076-456EA 1011.8 CAD
10076-456
Anti-DLL1 Rabbit Polyclonal Antibody
Antibodies
DLL1 antibody can be used in ELISA, and immunohistochemistry starting at 8 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR