Anti-PTPRN2 Chicken Polyclonal Antibody

49-631
10079-112EA 623.55 CAD
10079-112
Anti-PTPRN2 Chicken Polyclonal Antibody
IA-2 beta, a R8 receptor protein tyrosine phosphatase, is composed of a single transmembrane region and one putative tyrosine phosphatase catalytic domain. IA-2 beta is found within the secretory granule membrane of neuroendocrine cells and is a major antigen associated with type 1 diabetes, and the precursor of the 37 - kDa tryptic fragment that is found in insulin-dependent diabetes mellitus (IDDM) sera.

PTPRN2 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

PBS. No preservative added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTPRN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR