Anti-BMP5 Rabbit Polyclonal Antibody

Supplier: ProSci
49-649
10079-130EA 1011.8 CAD
10079-130
Anti-BMP5 Rabbit Polyclonal Antibody
Antibodies
BMP5 antibody can be used in ELISA, and immunohistochemistry starting at 2.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BMP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR