Anti-ARNT Rabbit Polyclonal Antibody

49-655
10079-142EA 811.46 CAD
10079-142
Anti-ARNT Rabbit Polyclonal Antibody
Transcriptional responses to many environmental and developmental stimuli are mediated by the basic helix-loop-helix (bHLH)-PAS family of proteins, which form homodimers that bind regulatory DNA sequences. ARNT/HIF-1 beta regulates the expression of different sets of genes in response to distinct environmental challenges. ARNT forms a complex with arylhydrocarbon receptor (AHR) to activate expression of genes encoding P450. Once AHR binds it ligand, it associates with ARNT for transport into the nucleus. ARNT also dimerizes with HIF-1alpha, -2 alpha or -3 alpha in response to low oxygen concentration to induce the expression of hypoxia-regulated genes encoding vascular endothelial growth and other genes. The expression of ARNT/HIF-1beta remains fairly constant and the protein is found in the nucleus and the cytoplasm.

ARNT antibody can be used in ELISA starting at 1:20000 - 1:100000, Western Blot starting at 1:500 - 1:5000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARNT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR