Anti-GDF2 Rabbit Polyclonal Antibody

49-670
10079-168EA 620.52 CAD
10079-168
Anti-GDF2 Rabbit Polyclonal Antibody
GDF2 antibody can be used in ELISA starting at 1:7500 - 1:30000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GDF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR