Anti-MLLT1 Rabbit Polyclonal Antibody

Supplier: ProSci

49-697
10079-220EA 955.07 CAD
10079-220
Anti-MLLT1 Rabbit Polyclonal Antibody
Antibodies
MLLT1 antibody can be used in ELISA starting at 0.5 ?g/mL, Western Blot starting at 1 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MLLT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR