Anti-MLLT4 Rabbit Polyclonal Antibody

Supplier: Prosci

49-700
10076-466EA 1011.8 CAD
10076-466
Anti-MLLT4 Rabbit Polyclonal Antibody
Antibodies
MLLT4 antibody can be used in ELISA starting at 1:16000 - 1:64000, Western Blot starting at 1:500 - 1:3000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MLLT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR