Anti-FOXO4 Rabbit Polyclonal Antibody

Supplier: Prosci
49-702
10076-470EA 955.07 CAD
10076-470
Anti-FOXO4 Rabbit Polyclonal Antibody
Antibodies
FOXO4 antibody can be used in ELISA starting at 1:20000 - 1:100000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:500.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR