Anti-LRRC3B Rabbit Polyclonal Antibody

Supplier: Prosci
49-709
10076-484EA 955.07 CAD
10076-484
Anti-LRRC3B Rabbit Polyclonal Antibody
Antibodies
LRRC3B antibody can be used in ELISA starting at 1:15000 - 1:60000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LRRC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR