Anti-MAGEB2 Rabbit Polyclonal Antibody

49-718
10079-230EA 811.46 CAD
10079-230
Anti-MAGEB2 Rabbit Polyclonal Antibody
MAGEB2 antibody can be used in ELISA starting at 1:1500 - 1:7000, Western Blot starting at 1:200 - 1:5000, and immunohistochemistry starting at 1.25 - 2.5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAGEB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR