Anti-PVR Rabbit Polyclonal Antibody

49-726
10079-246EA 736.85 CAD
10079-246
Anti-PVR Rabbit Polyclonal Antibody
CD155, a member of the immunoglobulin superfamily coding for a transmembrane protein has been shown to serve as an entry receptor for poliovirus, porcine pseudorabies virus, and bovine herpesvirus 1. CD155 gene is a transcriptional target of Sonic Hedgehog (SHH), a finding that has significance for the normal function of CD155 during development and the expression of CD155 in neuroectodermal tumors.

PVR antibody can be used in ELISA starting at 1:1000 - 1:6000, Western Blot starting at 1:200 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PVR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR