Anti-ST5 Rabbit Polyclonal Antibody

Supplier: Prosci

49-735
10079-262EA 954.81 CAD
10079-262
Anti-ST5 Rabbit Polyclonal Antibody
Antibodies
ST5 antibody can be used in Western Blot, and immunohistochemistry starting at 20 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ST5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR